Learn More
Biomatik Corporation Chicken Immunoglobulin A (IgA) ELISA Kit
Supplier:  Biomatik Corporation EKU10130
Encompass_Preferred
Chicken Immunoglobulin A (IgA) ELISA Kit for detecting IgA in serum, plasma, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 78-5,000pg/ml. Sensitivity: 32pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6656