Learn More
Biomatik Corporation Human Arachidonic Acid, AA ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC31179
Encompass_Preferred
The Human Arachidonic Acid, AA ELISA Kit allows for quantitative determination of Arachidonic Acid, AA in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human. Detection Range: 78.125 ng/mL-5000 ng/mL. Sensitivity: 69.748 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. Restriction: For Research Use Only.
Catalog No. 50-149-7403