Learn More
Biomatik Corporation High Sensitive Bovine Serum Albumin (BSA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08261
Encompass_Preferred
The High Sensitive Bovine Serum Albumin (BSA) ELISA Kit allows for quantitative determination of Bovine Serum Albumin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: All species. Detection Range: 246.9-20000 ng/mL. Sensitivity: 87.2 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. Cross Activity: No significant cross-reactivity or interference between Bovine Serum Albumin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4173