Learn More
Biomatik Corporation High Sensitive Human C Reactive Protein (CRP) ELISA Kit, 96 tests
Catalog No. 501504174
Encompass_Preferred
The High Sensitive Human C Reactive Protein (CRP) ELISA Kit allows for quantitative determination of C Reactive Protein in Serum, Plasma, Tissue homogenates, Cell lysates, Urine, Cerebrospinal fluid, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 62.5-4,000 pg/mL. Sensitivity: 23.3 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P02741. Cross Activity: No significant cross-reactivity or interference between C Reactive Protein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4174 Supplier Biomatik Corporation Supplier No. EKU08262
The High Sensitive Human C Reactive Protein (CRP) ELISA Kit allows for quantitative determination of C Reactive Protein in Serum, Plasma, Tissue homogenates, Cell lysates, Urine, Cerebrospinal fluid, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 62.5-4,000 pg/mL. Sensitivity: 23.3 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P02741. Cross Activity: No significant cross-reactivity or interference between C Reactive Protein and analogues was observed. Restriction: For Research Use Only.