Learn More
Biomatik Corporation High Sensitive Rat C Reactive Protein (CRP) ELISA Kit, 96 tests
Catalog No. 501504175
Encompass_Preferred
The High Sensitive Rat C Reactive Protein (CRP) ELISA Kit allows for quantitative determination of C Reactive Protein in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 15.62-1000 pg/mL. Sensitivity: 5.1 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P48199. Cross Activity: No significant cross-reactivity or interference between C Reactive Protein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4175 Supplier Biomatik Corporation Supplier No. EKU08263
The High Sensitive Rat C Reactive Protein (CRP) ELISA Kit allows for quantitative determination of C Reactive Protein in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 15.62-1000 pg/mL. Sensitivity: 5.1 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P48199. Cross Activity: No significant cross-reactivity or interference between C Reactive Protein and analogues was observed. Restriction: For Research Use Only.