Learn More
Biomatik Corporation High Sensitive Human Fibronectin (FN) ELISA Kit, 96 tests
Catalog No. 501504176
Encompass_Preferred
The High Sensitive Human Fibronectin (FN) ELISA Kit allows for quantitative determination of Fibronectin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.053 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P02751. Cross Activity: No significant cross-reactivity or interference between Fibronectin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4176 Supplier Biomatik Corporation Supplier No. EKU08265
The High Sensitive Human Fibronectin (FN) ELISA Kit allows for quantitative determination of Fibronectin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.053 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P02751. Cross Activity: No significant cross-reactivity or interference between Fibronectin and analogues was observed. Restriction: For Research Use Only.