Learn More
Biomatik Corporation High Sensitive Human Haptoglobin (Hpt) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08266
Encompass_Preferred
The High Sensitive Human Haptoglobin (Hpt) ELISA Kit allows for quantitative determination of Haptoglobin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.102 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P00738. Cross Activity: No significant cross-reactivity or interference between Haptoglobin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4177