Learn More
Biomatik Corporation High Sensitive Mouse Interleukin 10 (IL10) ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKU08273
The High Sensitive Mouse Interleukin 10 (IL10) ELISA Kit allows for quantitative determination interleukin 10 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 1.56-100 pg/mL. Sensitivity: 0.43 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P18893. Cross Activity: No significant cross-reactivity or interference between Interleukin 10 and analogues was observed. Restriction: For Research Use Only.