Learn More
Biomatik Corporation Human Complement Component 4d (C4d) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10279
Encompass_Preferred
Human Complement Component 4d (C4d) High-sensitive ELISA Kit for detecting C4d in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 9.4-600pg/ml. Sensitivity: 3.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P0C0L5. Gene ID: 100293534. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6686