Learn More
Biomatik Corporation High Sensitive Cattle Interleukin 6 (IL6) ELISA Kit, 96 tests
Catalog No. 501504179
Encompass_Preferred
The High Sensitive Cattle Interleukin 6 (IL6) ELISA Kit allows for quantitative determination interleukin 6 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 1.56-100 pg/mL. Sensitivity: 0.45 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P26892. Cross Activity: No significant cross-reactivity or interference between Interleukin 6 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4179 Supplier Biomatik Corporation Supplier No. EKU08275
The High Sensitive Cattle Interleukin 6 (IL6) ELISA Kit allows for quantitative determination interleukin 6 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 1.56-100 pg/mL. Sensitivity: 0.45 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P26892. Cross Activity: No significant cross-reactivity or interference between Interleukin 6 and analogues was observed. Restriction: For Research Use Only.