Learn More
Biomatik Corporation Pig Prolactin (PRL) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10313
Encompass_Preferred
Pig Prolactin (PRL) High-sensitive ELISA Kit for detecting PRL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 1.56-100ng/ml. Sensitivity: 0.61ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01238. Gene ID: 396965. Target Synonyms: LTH; Luteotropic Hormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6694