Learn More
Biomatik Corporation High Sensitive Human Nerve Growth Factor (NGF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08279
Encompass_Preferred
The High Sensitive Human Nerve Growth Factor (NGF) ELISA Kit allows for quantitative determination of Nerve Growth Factor in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 pg/mL. Sensitivity: 0.97 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P01138. Cross Activity: No significant cross-reactivity or interference between Nerve Growth Factor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4182