Learn More
Biomatik Corporation High Sensitive Human Tumor Necrosis Factor Alpha (TNFa) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08283
Encompass_Preferred
The High Sensitive Human Tumor Necrosis Factor Alpha (TNFa) ELISA Kit allows for quantitative determination of Tumor Necrosis Factor Alpha in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 pg/mL. Sensitivity: 0.52 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P01375. Cross Activity: No significant cross-reactivity or interference between Tumor Necrosis Factor Alpha and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4186