Learn More
Biomatik Corporation Human Homing Associated Cell Adhesion Molecule (HCAM) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08287
Encompass_Preferred
The Human Homing Associated Cell Adhesion Molecule (HCAM) ELISA Kit allows for quantitative determination of Homing Associated Cell Adhesion Molecule in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.56 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P16070. Cross Activity: No significant cross-reactivity or interference between Homing Associated Cell Adhesion Molecule and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4187