Learn More
Biomatik Corporation Rat Advanced Oxidation Protein Products (AOPP) ELISA Kit
Supplier:  Biomatik Corporation EKU10612
Encompass_Preferred
Rat Advanced Oxidation Protein Products (AOPP) ELISA Kit for detecting AOPP in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 24.69-2,000pg/ml. Sensitivity: 11.7pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6715