Learn More
Biomatik Corporation Mouse Alpha-1-Microglobulin (a1M) ELISA Kit
Supplier:  Biomatik Corporation EKU10623
Encompass_Preferred
Mouse Alpha-1-Microglobulin (a1M) ELISA Kit for detecting a1M in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 2.47-200ug/ml. Sensitivity: 0.94ug/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q07456. Gene ID: 11699. Target Synonyms: AMBP; UTI; HCP; EDC1; HI30; IATIL; ITILC; ITI; ITIL; Alpha 1 Microglobulin/Bikunin Precursor; Growth-inhibiting protein 19; Uristatin; Uronic-Acid-Rich Protein; Trypstatin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6720