Learn More
Biomatik Corporation 1, 25-Dihydroxyvitamin D3 (DHVD3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08306
Encompass_Preferred
The 1, 25-Dihydroxyvitamin D3 (DHVD3) ELISA Kit allows for quantitative determination of 1, 25-Dihydroxyvitamin D3 in Serum, Plasma and other biological fluids. Species Reactivity: All species. Detection Range: 123.5-10000 pg/mL. Sensitivity: 49.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between 1, 25-Dihydroxyvitamin D3 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4191