Learn More
Biomatik Corporation Hyaluronic Acid (HA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08324
Encompass_Preferred
Hyaluronic Acid (HA) ELISA Kit for detecting HA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 4.94-400ng/ml. Sensitivity: 1.71ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: Hyaluronan; Hyaluronate. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4193