Learn More
Biomatik Corporation Human Butyrophilin Subfamily 3, Member A1 (BTN3A1) ELISA Kit
Supplier:  Biomatik Corporation EKU10687
Encompass_Preferred
Human Butyrophilin Subfamily 3, Member A1 (BTN3A1) ELISA Kit for detecting BTN3A1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.124ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O00481. Gene ID: 11119. Target Synonyms: CD277; BT3.1; BTF5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6740