Learn More
Biomatik Corporation Human Collagen Type IV Alpha 5 (COL4a5) ELISA Kit
Supplier:  Biomatik Corporation EKU10736
Encompass_Preferred
Human Collagen Type IV Alpha 5 (COL4a5) ELISA Kit for detecting COL4a5 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.17ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P29400. Gene ID: 1287. Target Synonyms: COL4-A5; ASLN; ATS; CA54; Collagen Alpha-5 (IV) chain; Alport Syndrome. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6752