Learn More
Biomatik Corporation Cotinine (Cot) ELISA Kit
Supplier:  Biomatik Corporation EKU10751
Encompass_Preferred
Cotinine (Cot) ELISA Kit for detecting Cot in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 617.3-50,000pg/ml. Sensitivity: 233.7pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6756