Learn More
Biomatik Corporation Human Acidic Salivary Proline Rich Phosphoprotein 2 (PRH2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08365
Encompass_Preferred
The Human Acidic Salivary Proline Rich Phosphoprotein 2 (PRH2) ELISA Kit allows for quantitative determination of Acidic Salivary Proline Rich Phosphoprotein 2 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.55 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P02810. Cross Activity: No significant cross-reactivity or interference between Acidic Salivary Proline Rich Phosphoprotein 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4199