Learn More
Biomatik Corporation Rat Antithrombin (AT) ELISA Kit, 96 tests
Catalog No. 501504200
Encompass_Preferred
Rat Antithrombin (AT) ELISA Kit for detecting AT in plasma. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000ng/ml. Sensitivity: 6.9ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5M7T5. Gene ID: 304917. Target Synonyms: AT3; ATIII; SERPINC1; Anti-Thrombin Antibodies; Serpin Peptidase Inhibitor Clade C Member 1; Coding Sequence Signal Peptide Antithrombin Part 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4200 Supplier Biomatik Corporation Supplier No. EKU08366
Rat Antithrombin (AT) ELISA Kit for detecting AT in plasma. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000ng/ml. Sensitivity: 6.9ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5M7T5. Gene ID: 304917. Target Synonyms: AT3; ATIII; SERPINC1; Anti-Thrombin Antibodies; Serpin Peptidase Inhibitor Clade C Member 1; Coding Sequence Signal Peptide Antithrombin Part 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.