Learn More
Biomatik Corporation Human T-Cell Immunoreceptor With Ig And ITIM Domains Protein (TIGIT) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08370
Encompass_Preferred
Human T-Cell Immunoreceptor With Ig And ITIM Domains Protein (TIGIT) ELISA Kit for detecting TIGIT in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q495A1. Gene ID: 201633. Target Synonyms: VSIG9; VSTM3; V-Set And Immunoglobulin Domain Containing 9; V-Set And Transmembrane Domain Containing 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4201