Learn More
Biomatik Corporation Human Interleukin 2 Receptor Beta (IL2Rb) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08375
Encompass_Preferred
The Human Interleukin 2 Receptor Beta (IL2Rb) ELISA Kit allows for quantitative determination interleukin 2 Receptor Beta in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 78-5000 pg/mL. Sensitivity: 29 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P14784. Cross Activity: No significant cross-reactivity or interference between Interleukin 2 Receptor Beta and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4202