Learn More
Biomatik Corporation Human Protein Tyrosine Phosphatase Receptor Type Q (PTPRQ) ELISA Kit, 96 tests
Catalog No. 501504204
Encompass_Preferred
The Human Protein Tyrosine Phosphatase Receptor Type Q (PTPRQ) ELISA Kit allows for quantitative determination of Protein Tyrosine Phosphatase Receptor Type Q in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.058 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9UMZ3. Cross Activity: No significant cross-reactivity or interference between Protein Tyrosine Phosphatase Receptor Type Q and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4204 Supplier Biomatik Corporation Supplier No. EKU08386
The Human Protein Tyrosine Phosphatase Receptor Type Q (PTPRQ) ELISA Kit allows for quantitative determination of Protein Tyrosine Phosphatase Receptor Type Q in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.058 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9UMZ3. Cross Activity: No significant cross-reactivity or interference between Protein Tyrosine Phosphatase Receptor Type Q and analogues was observed. Restriction: For Research Use Only.