Learn More
Biomatik Corporation Rat Ghrelin (GHRL) ELISA Kit
Catalog No. 502286784
Encompass_Preferred
Rat Ghrelin (GHRL) ELISA Kit for detecting GHRL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78-5,000pg/ml. Sensitivity: 28pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9QYH7. Gene ID: 59301. Target Synonyms: MTLRP; Growth Hormone-Releasing Peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6784 Supplier Biomatik Corporation Supplier No. EKU10864
Rat Ghrelin (GHRL) ELISA Kit for detecting GHRL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78-5,000pg/ml. Sensitivity: 28pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9QYH7. Gene ID: 59301. Target Synonyms: MTLRP; Growth Hormone-Releasing Peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.