Learn More
Biomatik Corporation Human Farnesyl Diphosphate Synthase (FDPS) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08393
Encompass_Preferred
The Human Farnesyl Diphosphate Synthase (FDPS) ELISA Kit allows for quantitative determination of Farnesyl Diphosphate Synthase in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 62.5-4,000 pg/mL. Sensitivity: 26.3 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P14324. Cross Activity: No significant cross-reactivity or interference between Farnesyl Diphosphate Synthase and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4206