Learn More
Biomatik Corporation Human Glutathione S Transferase Mu 3, Brain (GSTm3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08396
Encompass_Preferred
The Human Glutathione S Transferase Mu 3, Brain (GSTm3) ELISA Kit allows for quantitative determination of Glutathione S Transferase Mu 3, Brain in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.15 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P21266. Cross Activity: No significant cross-reactivity or interference between Glutathione S Transferase Mu 3, Brain and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4208