Learn More
Biomatik Corporation Mouse Kelch Like ECH Associated Protein 1 (KEAP1) ELISA Kit, 96 tests
Catalog No. 501504210
Encompass_Preferred
Mouse Kelch Like ECH Associated Protein 1 (KEAP1) ELISA Kit for detecting KEAP1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Z2X8. Gene ID: 50868. Target Synonyms: INrf2; KLHL19; Cytosolic inhibitor of Nrf2; Kelch-like protein 19. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4210 Supplier Biomatik Corporation Supplier No. EKU08402
Mouse Kelch Like ECH Associated Protein 1 (KEAP1) ELISA Kit for detecting KEAP1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Z2X8. Gene ID: 50868. Target Synonyms: INrf2; KLHL19; Cytosolic inhibitor of Nrf2; Kelch-like protein 19. Restrictions: For Research Use Only. Not for use in diagnostic procedures.