Learn More
Biomatik Corporation Pig Anti-Mullerian Hormone (AMH) ELISA Kit, 96 tests
Catalog No. 501504211
Encompass_Preferred
Pig Anti-Mullerian Hormone (AMH) ELISA Kit for detecting AMH in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 1.88-30ng/ml. Sensitivity: 0.75ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P79295. Target Synonyms: MIF; MIH; MIS; Müllerian Inhibiting Factor; Müllerian Inhibiting Hormone; Müllerian Inhibiting Substance. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4211 Supplier Biomatik Corporation Supplier No. EKU08406
Pig Anti-Mullerian Hormone (AMH) ELISA Kit for detecting AMH in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 1.88-30ng/ml. Sensitivity: 0.75ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P79295. Target Synonyms: MIF; MIH; MIS; Müllerian Inhibiting Factor; Müllerian Inhibiting Hormone; Müllerian Inhibiting Substance. Restrictions: For Research Use Only. Not for use in diagnostic procedures.