Learn More
Biomatik Corporation Rat Leukemia Inhibitory Factor (LIF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08412
Encompass_Preferred
Rat Leukemia Inhibitory Factor (LIF) ELISA Kit for detecting LIF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P17777. Target Synonyms: CDF; D-FACTOR; HILDA; MLPLI; Cholinergic Differentiation Factor; Differentiation-Stimulating Factor; Melanoma-Derived LPL Inhibitor; Emfilermin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4214