Learn More
Biomatik Corporation Pig Laminin (LN) ELISA Kit
Supplier:  Biomatik Corporation EKU11034
Encompass_Preferred
Pig Laminin (LN) ELISA Kit for detecting LN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.78-50ng/ml. Sensitivity: 0.35ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: A0A287AG36; F1SAE9; F1S663. Target Synonyms: LAM. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6840