Learn More
Biomatik Corporation Liraglutide (LRT) ELISA Kit
Supplier:  Biomatik Corporation EKU11049
Encompass_Preferred
Liraglutide (LRT) ELISA Kit for detecting LRT in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 12.35-1,000pg/ml. Sensitivity: 4.64pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: Victoza. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6847