Learn More
Biomatik Corporation Mouse Carcinoembryonic Antigen Related Cell Adhesion Molecule 1 (CEACAM1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08434
Encompass_Preferred
Mouse Carcinoembryonic Antigen Related Cell Adhesion Molecule 1 (CEACAM1) ELISA Kit for detecting CEACAM1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5000pg/ml. Sensitivity: 32pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P31809. Gene ID: 26365. Target Synonyms: CD66a; BGP; BGP1; BGPI; CD66-A; Biliary Glycoprotein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4220