Learn More
Biomatik Corporation Rat Complement Component 2 (C2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08453
Encompass_Preferred
Rat Complement Component 2 (C2) ELISA Kit for detecting C2 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.19-12ng/ml. Sensitivity: 0.07ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6MG73. Gene ID: 24231. Target Synonyms: CO2; C3/C5 convertase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4222