Learn More
Biomatik Corporation Mouse Receptor Interacting Serine Threonine Kinase 3 (RIPK3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08458
Encompass_Preferred
Mouse Receptor Interacting Serine Threonine Kinase 3 (RIPK3) ELISA Kit for detecting RIPK3 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9QZL0. Gene ID: 56532. Target Synonyms: RIP3; RIP-like protein kinase 3; Receptor-interacting protein 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4223