Learn More
Biomatik Corporation Rat Insulin Like Growth Factor Binding Protein 7 (IGFBP7) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08464
Encompass_Preferred
Rat Insulin Like Growth Factor Binding Protein 7 (IGFBP7) ELISA Kit for detecting IGFBP7 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 62.5-4,000pg/ml. Sensitivity: 26.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: F1M9B2. Target Synonyms: FSTL2; IGFBP-7v; MAC25; PSF; TAF; IGFBP-rP1; MAC25 protein; Prostacyclin-stimulating factor; PGI2-stimulating factor; Tumor-derived adhesion factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4225