Learn More
Biomatik Corporation Rat Ectonucleotide Pyrophosphatase/Phosphodiesterase 2 (ENPP2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08473
Encompass_Preferred
Rat Ectonucleotide Pyrophosphatase/Phosphodiesterase 2 (ENPP2) ELISA Kit for detecting ENPP2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.064ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q64610. Gene ID: 84050. Target Synonyms: ATX; ATX-X; LysoPLD; NPP2; PD-IALPHA; PDNP2; Autotaxin; Extracellular lysophospholipase D. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4227