Learn More
Biomatik Corporation Multi-species Actin Alpha 2, Smooth Muscle (ACTa2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08517
Encompass_Preferred
Human/Mouse/Rat Actin Alpha 2, Smooth Muscle (ACTa2) ELISA Kit for detecting ACTa2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: 7.8-500ng/ml. Sensitivity: 2.9ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P62736. Gene ID: 59. Target Synonyms: ACT-A2; ACTSA; ACTVS; Asma; GIG46; Cell growth-inhibiting gene 46 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4237