Learn More
Biomatik Corporation Multi-species Actin Beta (ACTb) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08518
Encompass_Preferred
Human/Mouse/Rat Beta Actin (ACTB) ELISA Kit for detecting ACTB in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.55ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P60709. Gene ID: 60. Target Synonyms: Actin Beta; ACT-B; PS1TP5BP1; Actin, cytoplasmic 1; Actin, cytoplasmic 1, N-terminally processed. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4238