Learn More
Biomatik Corporation Human Protein Phosphatase, Mg2+/Mn2+ Dependent 1D (PPM1D) ELISA Kit
Supplier:  Biomatik Corporation EKU11378
Encompass_Preferred
Human Protein Phosphatase, Mg2+/Mn2+ Dependent 1D (PPM1D) ELISA Kit for detecting PPM1D in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O15297. Gene ID: 8493. Target Synonyms: PP2C-DELTA; WIP1; Wild-Type p53-Induced Phosphatase 1; Protein Phosphatase 2C,Delta Isoform; Protein Phosphatase 1D Magnesium-Dependent,Delta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6910