Learn More
Biomatik Corporation Mouse S100 Calcium Binding Protein A4 (S100A4) ELISA Kit
Supplier:  Biomatik Corporation EKU11413
Encompass_Preferred
Mouse S100 Calcium Binding Protein A4 (S100A4) ELISA Kit for detecting S100A4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P07091. Gene ID: 20198. Target Synonyms: S100-A4; MTS1; 18A2; 42A; CAPL; P9KA; PEL98; Calcium protein; Calvasculin; Metastasin; Protein Mts1; Placental calcium-binding protein; Murine placental homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6914