Learn More
Biomatik Corporation Cattle Haptoglobin (Hpt) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU08550
Encompass_Preferred
The Cattle Haptoglobin (Hpt) ELISA Kit allows for quantitative determination of Haptoglobin in Serum, Plasma and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 15.6-1000 ng/mL. Sensitivity: 5.9 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q2TBU0. Cross Activity: No significant cross-reactivity or interference between Haptoglobin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4241