Learn More
Biomatik Corporation Human Slit Homolog 3 (Slit3) ELISA Kit
Supplier:  Biomatik Corporation EKU11432
Encompass_Preferred
Human Slit Homolog 3 (Slit3) ELISA Kit for detecting Slit3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O75094. Gene ID: 6586. Target Synonyms: MEGF5; SLIL2; Multiple epidermal growth factor-like domains protein 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6919