Learn More
Biomatik Corporation Human SPARC Like Protein 1 (SPARCL1) ELISA Kit
Catalog No. 502286921
Encompass_Preferred
Human SPARC Like Protein 1 (SPARCL1) ELISA Kit for detecting SPARCL1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q14515. Gene ID: 8404. Target Synonyms: SC1; PIG33; Mast9; Hevin; High endothelial venule protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6921 Supplier Biomatik Corporation Supplier No. EKU11437
Human SPARC Like Protein 1 (SPARCL1) ELISA Kit for detecting SPARCL1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q14515. Gene ID: 8404. Target Synonyms: SC1; PIG33; Mast9; Hevin; High endothelial venule protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.