Learn More
Biomatik Corporation 1-Desamino 8D Arginine Vasopressin (DDAVP) ELISA Kit
Supplier:  Biomatik Corporation EKU02012
Encompass_Preferred
1-Desamino 8D Arginine Vasopressin (DDAVP) ELISA Kit for detecting DDAVP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 246.9-20,000pg/ml. Sensitivity: 90.9pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: d-DAVP; Stimate; Minirin; Desmopressin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5749