Learn More
Biomatik Corporation Rat Activating Transcription Factor 4 (ATF4) ELISA Kit
Supplier:  Biomatik Corporation EKU02107
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Rat. Detection Range: 78-5,000pg/mL. Sensitivity: 29pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9ES19. Target Synonyms: TAXREB67; TXREB; CREB-2; DNA-binding protein TAXREB67; Cyclic AMP-responsive element-binding protein 2; Tax-responsive enhancer element-binding protein 67. Restriction: For Research Use Only.
Catalog No. 50-228-5760