Learn More
Biomatik Corporation Rat Pepsinogen C (PGC) ELISA Kit, 96 tests
Catalog No. 501504248
Encompass_Preferred
Rat Pepsinogen C (PGC) ELISA Kit for detecting PGC in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.124ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04073. Gene ID: 24864. Target Synonyms: PG-C; Progastricsin; Gastricsin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4248 Supplier Biomatik Corporation Supplier No. EKU08577
Rat Pepsinogen C (PGC) ELISA Kit for detecting PGC in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.124ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04073. Gene ID: 24864. Target Synonyms: PG-C; Progastricsin; Gastricsin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.