Learn More
Biomatik Corporation Human Aldose Reductase Like Protein 1 (ARL1) ELISA Kit
Catalog No. 502285774
Encompass_Preferred
Human Aldose Reductase Like Protein 1 (ARL1) ELISA Kit for detecting ARL1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O60218. Gene ID: 57016. Target Synonyms: AKR1B10; AKR1B12; HIS; HSI; ALDRLn; Aldo-Keto Reductase Family 1,Member B10; Aldose Reductase-Related Protein; Small Intestine Reductase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5774 Supplier Biomatik Corporation Supplier No. EKU02236
Human Aldose Reductase Like Protein 1 (ARL1) ELISA Kit for detecting ARL1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O60218. Gene ID: 57016. Target Synonyms: AKR1B10; AKR1B12; HIS; HSI; ALDRLn; Aldo-Keto Reductase Family 1,Member B10; Aldose Reductase-Related Protein; Small Intestine Reductase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.